Anti-ZMIZ1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65767-100EA 11025 NOK
ABCAAB65767-100
Anti-ZMIZ1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ZMIZ1 (zinc finger, MIZ-type containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR