Anti-KCND3 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB65794-100EA 11400 NOK
ABCAAB65794-100
Anti-KCND3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KCND3 (potassium voltage-gated channel, Shal-related subfamily, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human