Anti-KCNJ2 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB65796-100EA 11725 NOK
ABCAAB65796-100
Anti-KCNJ2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KCNJ2 (potassium inwardly-rectifying channel, subfamily J, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse