Anti-NTF3 Rabbit polyclonal antibody unconjugated

ABCAAB65804-100EA 8125 NOK
ABCAAB65804-100
Anti-NTF3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: NTF3 (neurotrophin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR