Anti-AMD1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65820-100EA 8725 NOK
ABCAAB65820-100
Anti-AMD1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: AMD1 (adenosylmethionine decarboxylase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR