Anti-ATP5J Rabbit polyclonal antibody unconjugated

ABCAAB65841-100EA 7600 NOK
ABCAAB65841-100
Anti-ATP5J Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: ATP5J (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR