Anti-ANXA1 Rabbit polyclonal antibody unconjugated

ABCAAB65844-100EA 7775 NOK
ABCAAB65844-100
Anti-ANXA1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: ANXA1 (annexin A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR