Anti-MIF Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65869-100EA 11975 NOK
ABCAAB65869-100
Anti-MIF Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MIF (macrophage migration inhibitory factor (glycosylation-inhibiting factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR