Anti-BNIP3 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65874-100EA 11725 NOK
ABCAAB65874-100
Anti-BNIP3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BNIP3 (BCL2/adenovirus E1B 19kDa interacting protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR