Anti-MASP3 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB65900-100EA 8050 NOK
ABCAAB65900-100
Anti-MASP3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MASP3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: catalytic domain
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR