Anti-CD14 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: MEM-15]

ABCAAB65950-50EA 7550 NOK
ABCAAB65950-50
Anti-CD14 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: MEM-15]
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: monoclonal
Clone: MEM-15
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR