Anti-MTR Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB66039-100EA 11850 NOK
ABCAAB66039-100
Anti-MTR Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MTR (5-methyltetrahydrofolate-homocysteine methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR