Anti-Proteinase Activated Receptor 3 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB66068-100EA 10825 NOK
ABCAAB66068-100
Anti-Proteinase Activated Receptor 3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Proteinase Activated Receptor 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human