Anti-TIMM13 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB66142-50EA 8450 NOK
ABCAAB66142-50
Anti-TIMM13 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TIMM13 (translocase of inner mitochondrial membrane 13 homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human