Anti-Influenza A Virus Nucleoprotein Mouse Monoclonal Antibody [clone: B247M]
Leverandør: Abcam
ABCAAB66191-500EA 8350 NOK
ABCAAB66191-500
Anti-Influenza A Virus Nucleoprotein Mouse Monoclonal Antibody [clone: B247M]
Antistoffer
Type: Primary
Antigen: Influenza A Virus Nucleoprotein
Clonality: monoclonal
Clone: B247M
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå