Anti-LAMTOR5 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB66258-50EA 8500 NOK
ABCAAB66258-50
Anti-LAMTOR5 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LAMTOR5 (late endosomal/lysosomal adaptor, MAPK and MTOR activator 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human