Anti-APOM Mouse monoclonal antibody unconjugated [clone: 8F12C6B8]

Leverandør: Abcam
ABCAAB66379-100EA 12175 NOK
ABCAAB66379-100
Anti-APOM Mouse monoclonal antibody unconjugated [clone: 8F12C6B8]
Antistoffer
Type: Primary
Antigen: APOM (apolipoprotein M)
Clonality: monoclonal
Clone: 8F12C6B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR