Anti-ECSIT Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB66380-100EA 8425 NOK
ABCAAB66380-100
Anti-ECSIT Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ECSIT (ECSIT signalling integrator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR