Anti-DLX3 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB66390-100EA 8550 NOK
ABCAAB66390-100
Anti-DLX3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DLX3 (distal-less homeobox 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR