Anti-GABRB3 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB66430-100EA 11475 NOK
ABCAAB66430-100
Anti-GABRB3 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: GABRB3 (gamma-aminobutyric acid (GABA) A receptor, beta 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR