Anti-GRIK2 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB66440-100EA 11475 NOK
ABCAAB66440-100
Anti-GRIK2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GRIK2 (glutamate receptor, ionotropic, kainate 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human