Anti-CLCNKB Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB66460-200EA 8350 NOK
ABCAAB66460-200
Anti-CLCNKB Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CLCNKB (chloride channel, voltage-sensitive Kb)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR