Anti-SDHA Rabbit polyclonal antibody unconjugated

ABCAAB66484-100EA 7600 NOK
ABCAAB66484-100
Anti-SDHA Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: SDHA (succinate dehydrogenase complex, subunit A, flavoprotein (Fp))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR