Anti-PBX1 Mouse Monoclonal Antibody [clone: 146C1a]

Leverandør: Abcam
ABCAAB66496-100EA 8500 NOK
ABCAAB66496-100
Anti-PBX1 Mouse Monoclonal Antibody [clone: 146C1a]
Antistoffer
Type: Primary
Antigen: PBX1 (pre-B-cell leukemia homeobox 1)
Clonality: monoclonal
Clone: 146C1a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR