Anti-CHD5 Rabbit polyclonal antibody unconjugated

ABCAAB66516-100EA 7775 NOK
ABCAAB66516-100
Anti-CHD5 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CHD5 (chromodomain helicase DNA binding protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR