Anti-GTPBP2 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB66519-100EA 8350 NOK
ABCAAB66519-100
Anti-GTPBP2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GTPBP2 (GTP binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR