Anti-TFPI Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB66544-100EA 10925 NOK
ABCAAB66544-100
Anti-TFPI Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TFPI (tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR