Anti-KCND2 Rabbit polyclonal antibody unconjugated

ABCAAB66639-100EA 7600 NOK
ABCAAB66639-100
Anti-KCND2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: KCND2 (potassium voltage-gated channel, Shal-related subfamily, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR