Anti-TOX4 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB66651-100EA 7950 NOK
ABCAAB66651-100
Anti-TOX4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TOX4 (TOX high mobility group box family member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR