Anti-APCS Rabbit polyclonal antibody unconjugated

ABCAAB66715-100EA 7425 NOK
ABCAAB66715-100
Anti-APCS Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: APCS (amyloid P component, serum)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR