Anti-KCNIP2 Rabbit polyclonal antibody unconjugated

ABCAAB66741-200EA 7725 NOK
ABCAAB66741-200
Anti-KCNIP2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: KCNIP2 (Kv channel interacting protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR