Anti-OPN3 Rabbit polyclonal antibody unconjugated

ABCAAB66742-100EA 7550 NOK
ABCAAB66742-100
Anti-OPN3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: OPN3 (opsin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR