Anti-ASF1A Rabbit polyclonal antibody unconjugated

ABCAAB66754-100EA 7250 NOK
ABCAAB66754-100
Anti-ASF1A Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: ASF1A (anti-silencing function 1A histone chaperone)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR