Anti-C5b-9 Mouse monoclonal antibody unconjugated [clone: aE11]

Leverandør: Abcam
ABCAAB66768-100EA 11925 NOK
ABCAAB66768-100
Anti-C5b-9 Mouse monoclonal antibody unconjugated [clone: aE11]
Antistoffer
Type: Primary
Antigen: C5b-9
Clonality: monoclonal
Clone: aE11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR