Anti-HBEGF Mouse monoclonal antibody unconjugated [clone: 4G10]
Leverandør: Abcam
ABCAAB66792-100EA 11775 NOK
ABCAAB66792-100
Anti-HBEGF Mouse monoclonal antibody unconjugated [clone: 4G10]
Antistoffer
Type: Primary
Antigen: HBEGF (heparin-binding EGF-like growth factor)
Clonality: monoclonal
Clone: 4G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human