Anti-C5 / C5a Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB66854-100EA 12875 NOK
ABCAAB66854-100
Anti-C5 / C5a Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: C5 / C5a
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR