Anti-BTF3 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB66940-100EA 8625 NOK
ABCAAB66940-100
Anti-BTF3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BTF3 (basic transcription factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR