Anti-HIV1 p15 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB66951-100EA 10900 NOK
ABCAAB66951-100
Anti-HIV1 p15 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: HIV1 p15
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå