Anti-NSMCE1 Rabbit polyclonal antibody unconjugated

ABCAAB66956-100EA 7600 NOK
ABCAAB66956-100
Anti-NSMCE1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: NSMCE1 (non-SMC element 1 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR