Anti-DDX11 Mouse polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB66971-50EA 11475 NOK
ABCAAB66971-50
Anti-DDX11 Mouse polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: DDX11 (DEAD/H (Asp-Glu-Ala-Asp/His) box helicase 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human