Anti-LMAN2 Mouse Monoclonal Antibody [clone: 264C4a]

Leverandør: Abcam
ABCAAB67000-100EA 11600 NOK
ABCAAB67000-100
Anti-LMAN2 Mouse Monoclonal Antibody [clone: 264C4a]
Antistoffer
Type: Primary
Antigen: LMAN2 (lectin, mannose-binding 2)
Clonality: monoclonal
Clone: 264C4a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR