Anti-NCOA1 Mouse monoclonal antibody unconjugated [clone: 3532C6a]

ABCAAB67101-100EA 7600 NOK
ABCAAB67101-100
Anti-NCOA1 Mouse monoclonal antibody unconjugated [clone: 3532C6a]
Type: Primary
Antigen: NCOA1 (nuclear receptor coactivator 1)
Clonality: monoclonal
Clone: 3532C6a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR