Anti-KATNA1 Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB67206-50EA 11650 NOK
ABCAAB67206-50
Anti-KATNA1 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KATNA1 (katanin p60 (ATPase containing) subunit A 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR