Anti-ARHGEF17 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB67278-100EA 11650 NOK
ABCAAB67278-100
Anti-ARHGEF17 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ARHGEF17 (Rho guanine nucleotide exchange factor (GEF) 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human