Anti-SIRT2 Rabbit polyclonal antibody unconjugated
ABCAAB67299-100EA 7775 NOK
ABCAAB67299-100
Anti-SIRT2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: SIRT2 (sirtuin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat