Anti-GRIK1 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB67316-100EA 11725 NOK
ABCAAB67316-100
Anti-GRIK1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GRIK1 (glutamate receptor, ionotropic, kainate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse