Anti-LGALS3BP Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67353-50EA 11775 NOK
ABCAAB67353-50
Anti-LGALS3BP Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LGALS3BP (lectin, galactoside-binding, soluble, 3 binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human