Anti-Protein C inhibitor Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67368-50EA 8625 NOK
ABCAAB67368-50
Anti-Protein C inhibitor Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Protein C inhibitor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human