Anti-POLE Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67420-50EA 11475 NOK
ABCAAB67420-50
Anti-POLE Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: POLE (polymerase (DNA directed), epsilon, catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human