Anti-IGF2BP2 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67434-50EA 8625 NOK
ABCAAB67434-50
Anti-IGF2BP2 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IGF2BP2 (insulin-like growth factor 2 mRNA binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human